Would you like to log in again?
DB-1 Columns, Agilent Technologies
Supplier: Agilent
J&W
123-1064 125-1015E 125-1011 124-1034 125-10HB 123-1013 125-1005 123-1034E 127-1046E 125-1017 123-1065-INT 125-1025 122-106E 127-1046 127-1013E 123-1033-INT 123-1032E 123-1012 123-106E 126-1012 125-1011E 123-1061 123-105C 125-1039 123-105C 125-1002E 125-10B5 123-1064 125-1065 125-103K 125-1062 125-1012 125-106J 121-1022 128-1052 125-1035 127-1012 123-102F 123-1065 123-1034 125-1037 123-1063 123-1055 122-1063 125-1035 123-103E 127-1013 123-1011 123-1034 125-1032 127-1023 123-1031 G3903-61004 125-1015E 125-106J 123-1065 128-1012 123-1011 123-1056 125-1014 125-1034E 125-100B 126-1012 125-103J 125-1064 125-1062 125-1055 125-100B 128-1012 125-1012 122-1012 125-1034 125-1034 122-106E 123-1032 122-1033 122-1032 125-101K 125-1005 123-106G 125-10HB 123-106B 123-1063E 12A-1015 125-1035E 123-1031 123-1013 125-101J 125-102J 122-1033 123-1061 123-1033 12A-1015 125-1064 123-103B 125-1017 125-10H5 125-103J 122-1063 123-1065E 123-1015 123-1014 125-1015 125-1037 123-1055 123-1065-INT 125-103KE 121-1022 123-1033 122-1062 128-1022 123-1015 123-1033E 126-1013 125-106JE 123-105F 123-1056 123-1062 125-1032 123-106G 123-1033E 123-1022 125-102J 125-1039 121-1023 123-102F 123-1032E 124-1032 127-100A 125-101K 123-1035 122-1012 123-1026 122-1062 125-1011 127-1013 124-1032 122-1032 127-1012E 123-1065E 127-100A 125-103KE 125-1034E 123-1032-INT 125-101J 123-1062 123-1022 123-103B 125-1065 123-1063E 128-1022 125-1025 125-1002 123-1014 123-1034E 123-105F 123-1027 125-1035E 127-1012 123-1035 123-106E 123-1026 122-1013 127-1023 125-1002 123-1063 123-1033-INT 127-1046 127-1012E 125-103K 123-103E 125-10B5 125-1015 123-1032-INT 127-1046E 122-1013 125-103B 125-1014 124-1034 123-106B 127-1043 121-1023 127-1043 128-1052 125-10H5 125-106JE 123-1027 127-1013E 125-1055 123-1032 123-1012 127-1022 125-103B 125-1011E
AG128-1052EA 1019.13 USD InStock
CAAG127-100A CAAG127-1013 CAAG127-1012 CA122-1062 CAAG127-1012E CA122-1063 CAAG125-1002 CAAG125-1005 CA122-106E AG128-1052 CAAG125-100B AG123-1034E CAAG123-1032E AG12A-1015 CAAG125-1014 CAAG1231033INT CA125-101J AG123-106B AG123-106E AG128-1022 AG123-1033E CAAG123-1065E AG126-1012 AG122-1013 AG126-1013 AG122-1012 AG123-106G CAAG127-1046E CA123-1033E CA125-1015 AG123-1032E CAAG127-1023 CA125-1012 CA125-1011 CAAG125-1039 CAAG125-1037 AG127-1012E CAAG125-103B AG125-1062 AG125-1065 AG125-1064 CAAG124-1034 CAAG124-1032 CA123-1032 CAAG125-103K CA123-1035 CA123-1033 CA123-1034 AG121-1022 AG121-1023 CA123-103B AG127-1013E AG125-1055 AG128-1012 CA122-1033 CA122-1032 AG127-1012 CAAG125-1017 AG127-1013 AG125-10HB CAAG125-10B5 AG125-103KE AG127-100A AG123-1065E CAAG125-101K CA123-1013 AG125-10H5 CA123-1014 CA123-1012 CAAG125-1025 CA123-1015 AG127-1023 AG125-106J AG127-1022 CAAG126-1012 CAAG125-102J CA125-1035E CAAG123-1034E CAAG123-1061 AG125-10B5 AG125-1002E AG125-101J AG123-1033-INT CAAG125-10H5 AG125-101K CAAG125-1034E AG125-1015 AG125-1014 CAAG123-105C CAAG125-10HB CAAG123-105F AG125-1017 CAAG128-1022 AG123-1063E AG125-1015E AG127-1046 AG125-100B AG127-1043 AG123-1012 AG123-1011 AG123-1014 AG123-1013 AG125-1012 AG123-1015 AG125-1011 AG125-1005 AG125-1002 CAAG123-106B CAAG125-106JE CAAG123-106E CAAG123-106G AG125-103K AG125-103J CAAG123-103E CAAG12A-1015 AG127-1046E AG125-1037 AG125-1035 AG125-1039 CAAG125-1064 CAAG125-1062 AG125-103B AG125-102J CAAG123-1056 AG123-1065-INT CAAG123-1055 AG125-1034 AG125-1032 CAAG125-1011E AG125-1025 CAAG125-106J CA123-1064 CA123-1065 CA123-1062 CA122-1013 CA122-1012 CA123-1063 CA128-1012 CA125-103J AG125-1034E CAAG123-1022 CAAG123-1063E AGG3903-61004 AG122-1033 AG122-1032 CA121-1022 CA121-1023 AG125-1035E CAAG123-1031 AG125-1011E AG123-105C AG123-1056 AG123-1055 CAAG123-102F CAAG1231065INT CAAG127-1046 AG123-1061 CA125-1035 AG123-1063 CAAG127-1043 AG123-1062 AG123-1065 AG123-1064 CAAG128-1052 AG123-105F CAAG123-1027 CAAG123-1026 CA125-1034 AG123-1032-INT CA125-1032 AG124-1034 AG124-1032 AG122-1062 AG123-1022 AG122-1063 AG123-1027 AG123-1026 AG123-1032 AG123-1031 CAAG125-103KE AG123-102F CA125-1065 CAAG1231032INT CAAG125-1015E AG122-106E AG123-103B CAAG123-1011 AG123-1034 AG123-1033 AG123-1035 AG125-106JE CAAG127-1013E AG123-103E CA125-1055
DB-1 Columns, Agilent Technologies
Chromatography Columns
A reliable, crosslinked, nonpolar column with low bleed and a high temperature limit, delivering superb performance for general purpose applications.
  • 100% Dimethylpolysiloxane
  • Nonpolar and low bleed with a high temperature limit
  • Wide range of applications
  • Bonded, crosslinked, and solvent rinsible
  • Equivalent to USP Phase G2
  • Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1
  • Available with DuraGuard (GC column with a built-in guard column)
  • Wide range of column dimensions available
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-1 is nonpolar and low-bleed and operates under high temperature limits. It is an excellent choice for general purpose use in a wide variety of applications. Precision-engineered DB-1 is bonded, crosslinked and solvent rinsible, and is equivalent to USP phase G2.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty