Would you like to log in again?
DB-1ms Columns, Agilent Technologies
Supplier: Agilent
123-0112 128-0112 122-0132 123-0162 127-0122 127-0123 127-0112LTM 122-0162 122-0132 123-0112 128-0122 122-0112 123-0132 127-0122 G3900-65002 123-0132 123-0131 122-0162 123-0162 128-0112 128-0122 122-0112 G3900-65002 127-0112LTM 127-0112 127-0112 123-0131
AG127-0122EA 831.77 USD InStock
AG127-0122 AG127-0123 CAAG123-0131 AG123-0132 CAAG1270112LTM AG123-0131 AG123-0112 AG128-0112 CAAG127-0122 CA122-0112 CA122-0132 AG123-0162 AGG3900-65002 CA128-0112 CA127-0112 AG127-0112 CAAG123-0162 CAG3900-65002 AG127-0112LTM AG122-0162 AG128-0122 AG122-0112 AG122-0132 CAAG123-0112 CA123-0132 CA122-0162 CA128-0122
DB-1ms Columns, Agilent Technologies
Chromatography Columns
An excellent general-purpose GC column, ideal for GC/MS of active analytes commonly found in drug screening and environmental analyses.
  • 100% Dimethylpolysiloxane
  • Identical selectivity to DB-1
  • Nonpolar
  • Very low bleed characteristics, ideal for GC/MS
  • Improved acid performance compared to standard 100% dimethylpolysiloxane columns
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Bonded, crosslinked, and solvent rinsible
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1, ZB-1ms
Agilent J&W DB-1ms is a nonpolar, low-bleed column, with an extended upper temperature limit and excellent inertness for acids and bases. The low-bleed stationary phase reduces column contribution to background noise, resulting in a higher signal-to-noise ratio and better sensitivity for improved mass-spectral purity and accurate library identification.
Bonded and crosslinked, this highly inert column provides maximum response for active analytes commonly found in drug screening and environmental analyses.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty