Would you like to log in again?
DB-5ms Columns, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
128-5522 122-5522 122-5532-INT 123-5563 123-5526 123-5531 128-5552 123-5513 125-5512 123-5536 128-5552 122-5513 122-5533E 123-5536 122-5563 122-5531 125-553J 123-5511 122-5511 123-5531 122-5562E 123-5566 122-5533E 122-5552 125-553J 123-5566 121-5522 122-5522 125-5537 122-5512LTM 122-5532E 122-5512 125-5512 122-5561 123-5513 128-5512 125-5532 123-5512 123-5532E 125-5532 123-5562 122-5511 122-5563 123-5562 122-5532E 122-5516 123-5533 122-5512LTM 122-5561 122-5532 122-5562 122-5516-INT 123-5563 121-5522 122-5516-INT 123-5512 123-5533 123-5526 122-5536 122-5512 122-5533 122-5536 122-5531 123-5511 125-5537 123-5532E 122-5513 128-5512 123-5532 122-5562E 122-5516 122-5532-INT 122-5562 122-5533 123-5532 122-5552 122-5532 128-5522
AG125-553JEA 934 USD InStock
AG125-553J AG128-5522 AG122-5563 CA76514-546 AG122-5562 AG122-5533E AG123-5566 AG122-5561 CAAG122-5533E CA76514-542 AG123-5526 AG122-5516 AG122-5513 AG123-5531 AG122-5512 AG122-5511 AG125-5537 AG122-5532-INT AG122-5512LTM CAAG123-5511 CAAG122-5522 CAAG122-5532E AG123-5533 AG125-5532 AG123-5532 AG122-5562E AG123-5536 AG122-5516-INT CA122-5516 CA123-5563 CAAG1225512LTM CA122-5513 AG122-5522 CA128-5512 CAAG1225516INT CAAG123-5562 CAAG123-5566 CAAG125-5537 CAAG122-5511 AG123-5532E CAAG125-553J AG122-5536 CAAG123-5532E AG125-5512 AG122-5533 AG122-5532 AG122-5531 CA123-5536 CA128-5522 CA122-5562 CA122-5561 CAAG123-5513 CAAG123-5512 CAAG122-5563 CA123-5532 CA123-5533 AG121-5522 CA121-5522 CAAG1225532INT CAAG123-5531 CAAG122-5562E AG123-5513 AG122-5552 AG123-5512 AG128-5512 AG123-5511 AG128-5552 AG122-5532E CA122-5536 CAAG128-5552 AG123-5563 CA122-5533 AG123-5562 CA122-5531 CAAG123-5526 CAAG125-5512 CA125-5532 CAAG122-5552
DB-5ms Columns, Agilent Technologies
Chromatography Columns
A nonpolar column with high inertness and improved signal-to-noise for analysis of aromatics and semi volatiles.
  • Phenyl arylene polymer virtually equivalent to (5%-phenyl)-methylpolysiloxane
  • Nonpolar with very low bleed characteristics, ideal for GC/MS
  • Excellent inertness for active compounds
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Bonded, crosslinked, and solvent rinsible
  • Exact replacement of HP-5TA
  • Close equivalent to USP phase G27
  • Test mix available
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency Performance Summary shipped with each column
Agilent J&W DB-5ms is a nonpolar phenyl arylene polymer, virtually equivalent to (5%-phenyl)-methylpolysiloxane, featuring an excellent signal-to-noise ratio for analytical applications. This improved performance gives better sensitivity and mass spectral integrity and delivers excellent inertness with active compounds.
This column is a close equivalent to USP phase G27, and is very low bleed, making it ideal for GC/MS.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty