Would you like to log in again?
CrossLab Dissolution Shafts, Agilent Technologies
Supplier: Agilent
CrossLab
13-3630 14-3613 13-3629 14-3636 13-3614 13-3629 13-3614 13-3622 14-3612 14-3634 14-3631 16-3613 13-3630 14-3620 13-3615 14-3620 13-3632 14-3624 14-3624 13-3615 14-3634 16-3631 16-3631 13-3622 14-3613 13-3632 14-3612 14-3631 16-3613
AG13-3615EA 295.16 USD InStock
CAAG13-3629 AG13-3615 CAAG14-3631 CAAG13-3622 AG13-3614 AG14-3620 CAAG14-3613 CAAG14-3612 CAAG14-3634 AG13-3632 AG14-3624 AG13-3630 AG16-3613 AG16-3631 CAAG13-3615 AG13-3629 CAAG13-3614 CAAG16-3613 CAAG14-3620 AG13-3622 AG14-3631 CAAG13-3632 CAAG14-3624 CAAG16-3631 CAAG13-3630 AG14-3636 AG14-3613 AG14-3634 AG14-3612
CrossLab Dissolution Shafts, Agilent Technologies
Dissolution Apparatus
Shafts of different materials and lengths, individually serialized to meet standard USP design.
  • Electropolished stainless steel, PTFE-coated, gold-plated or mini basket shafts available
  • Individually serialized
  • Three-clip shafts to meet standard USP design
  • O-ring shafts for automated systems
  • Conical shafts to support air bubble dispersion
  • A range of basket shaft lengths are available to accommodate various models and volume configurations
Agilent provides stainless steel, PTFE-coated, gold-plated, or mini shafts that are individually serialized. Several basket and paddle shaft lengths and options are available to meet standard USP design.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty