Copper, stainless steel, and UltiMetal Plus tubing for GC applications.
- Copper tubing can be used for most applications, except those requiring the highest purity gas delivery system
- Stainless steel and UltiMetal Plus tubing are appropriate for more demanding applications
Agilent GC tubing includes high-quality copper, stainless steel, and UltiMetal Plus tubing, which are manufactured and cleaned for chromatographic use. Agilent tubing comes in different lengths and depending on your flow requirements, is available in ¹/₁₆, ¹/₈, or ¹/₄". UltiMetal Plus tubing is available in ¹/₁₆" for inert environments.