Would you like to log in again?
Bond Elut DEA, Agilent Technologies
Supplier: Agilent Technologies
Bond Elut™
14102043 14102043
AG14102043PK 255 USD InStock
AG14102043 CAAG14102043
Bond Elut DEA, Agilent Technologies
Chromatography Columns
Bond Elut DEA is ideal for aqueous anion-exchange or as a polar sorbent with nonpolar matrices.
  • Weak anion exchanger
  • More polar than C8 but less polar than C2 or CN-E
  • Alkyl side chains exhibit moderately nonpolar characteristics
Bond Elut DEA (diethylaminopropyl) is similar to Bond Elut NH2, but with a slightly lower capacity as an anion-exchange sorbent. DEA has a moderately nonpolar character due to the alkyl side chains on the amino functionality. These groups still afford a medium level of polarity, higher than C8, but less polar than C2 or CN-E.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty