Bond Elut SPE cartridges are available in a range of chemistries, particle sizes, and bed masses.
- Cartridges available in sizes from 1 to 150 ml and in a range of chemistries, sorbent particle sizes, and bed masses
- Bond Elut Jr is Luer-compatible
- Large reservoir capacity (LRC) tubes offer flexibility
- Also available for special order requests
Bond Elut SPE cartridges range from 1 to 150 ml in a wide range of bonded silica and polymeric chemistries, sorbent particle sizes, and bed masses. The Luer-compatible Bond Elut Jr and the large reservoir capacity (LRC) tubes offer flexibility across a range of sorbent bed masses. Tabless (flangeless) versions of the straight barrel cartridges are also available.