Would you like to log in again?
Crosslab DB-HT Simulated Distillation Columns (moved), Agilent Technologies
Supplier: Agilent Technologies
145-1009 145-1009 145-1001 145-1001
AG145-1001EA 391 USD InStock
AG145-1001 CA145-1001 CA145-1009 AG145-1009
Crosslab DB-HT Simulated Distillation Columns (moved), Agilent Technologies
Chromatography Columns
Low-bleed GC columns with a 'boiling point' phase for high-temperature simulated distillation.
  • 100% Dimethylpolysiloxane
  • 'Boiling point'mphase for high-temperature simulated distillation
  • Durable stainless steel tubing
  • 430 °C upper temperature limit
  • Distillation range of C6 to C110+
  • Low bleed, even at 430 °C
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column"
Agilent J&W DB-HT Simulated Distillation columns feature a 'boiling point' phase for high-temperature simulated distillation, and durable stainless steel tubing for consistently excellent performance. These low-bleed columns have a distillation range of C6 to C110 with an operating temperature range of –60 to 430 °C. The columns are bonded, crosslinked, and solvent rinsable, and are rigorously tested to some of the strictest industry QC standards to ensure accurate results.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty