Would you like to log in again?
CrossLab Autosampler Syringes and Other Supplies, Agilent Technologies
Supplier: Agilent
CrossLab
1510146800 4710003200 4710003200 5410046300 5410046300 9910116800 6610012100 9910116800 4710002300 1510146800 4710002300 210131490 210131490 6610012100
AG5410046300EA 286.96 USD InStock
AG5410046300 CAAG4710003200 CAAG1510146800 CAAG4710002300 AG6610012100 AG9910116800 CAAG210131490 AG210131490 CAAG5410046300 AG1510146800 AG4710003200 CAAG9910116800 AG4710002300 CAAG6610012100
CrossLab Autosampler Syringes and Other Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Dispensing syringe and replacement plungersfor use with PSD 100/120 Programmable Sample Dispenser.
  • Solutions are drawn into and held in the dispensing capillary by the action of the syringe
  • All solutions are separated by an air gap in the dispensing capillary to prevent precipitation or other chemical reactions during sampling
  • The collected solutions are then dispensed into the graphite tube located in the furnace workhead
  • PTFE tipped plunger provides good sealing, reducing bubble formation and preventing cross contamination
  • 100 µl syringe provides dispensing range from 1 to 70 microlitres with minimum injection volume 1 microlitre and <1 % repeatability of dispensed volume
  • Rinsing of the dispensing probe is completed between collection for each solution component and after every injection
Agilent provides a range of supplies for the PSD 100/120 Programmable Sample Dspenser used with Agilent GFAAS systems, including sample vials, beakers, adapters, dispensing capillary assemblies, dispensing syringe and replacement plungers.​
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty