Would you like to log in again?
CrossLab Dissolution Paddles, Agilent Technologies
Supplier: Agilent
CrossLab
14-3603 14-3594 14-3608 16-3603 13-3592 13-3593 14-3597 13-3592 13-3596 14-3600 14-3605 13-3593 16-3602 14-3606 14-3597 14-3599 14-3595 16-3602 14-3603 14-3607 14-3590 14-3590 16-3603 14-3599 14-3602 14-3604 14-3606 14-3595 14-3600 14-3607 14-3604 14-3608 14-3594 14-3605 14-3602 13-3596
AG14-3590EA 449.67 USD InStock
CA14-3603 AG14-3590 CAAG14-3594 AG14-3600 CAAG16-3602 CAAG14-3597 CAAG16-3603 CAAG14-3595 AG14-3604 AG14-3603 AG13-3596 AG14-3602 CAAG14-3599 AG14-3608 AG14-3607 AG13-3592 AG14-3606 AG13-3593 AG14-3605 CAAG14-3608 CAAG14-3607 CAAG14-3590 AG14-3595 AG14-3594 AG16-3603 AG14-3599 AG14-3597 CAAG14-3602 CAAG13-3596 CAAG14-3600 AG16-3602 CAAG13-3592 CAAG14-3606 CAAG14-3605 CAAG13-3593 CAAG14-3604
CrossLab Dissolution Paddles, Agilent Technologies
Dissolution Apparatus
Available in stainless steel, PTFE-coated, or PEEK in a range of sizes.
  • All paddles are serialized and designed to meet USP specifications for Apparatus 2
  • Select from electropolished stainless steel, PTFE-coated, or the biocompatible, chemically inert PEEK paddles in a range of sizes
  • Additionally, mini and mega paddles for small- and large-volume dissolution testing are available
All paddles are serialized and designed to meet USP specifications for USP Apparatus 2 including an individual CoC as standard. Paddles are available in stainless steel, PTFE-coated, or PEEK in a range of standard, mega, or mini sizes.
Certifications: Each paddle includes an individual Certificate of Conformance as standard.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty