Would you like to log in again?
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: Agilent
CrossLab
17-4045 17-4010 17-4021 K1200-00855 17-4020 17-4004 17-4140 17-4003 17-4015 17-4050 17-4003 17-4010 17-4045 K1200-00855 17-4130 17-4040 17-4001 17-4055 17-4140 17-4110 17-4120 17-4000 17-4021 17-4025 17-4130 17-4005 17-4120 17-4011 17-4011 17-4055 17-4015 17-4100 17-4001 17-4005 17-4100 17-4025 17-4004 17-4050 17-4040 17-4020 17-4000 17-4110
AG17-4140PK 144.92 USD InStock
AG17-4140 CAAG17-4050 AG17-4020 AG17-4040 AG17-4045 CAAG17-4010 AG17-4001 AG17-4100 CAAG17-4055 CAAG17-4011 CAAG17-4110 AG17-4120 AG17-4021 AGK1200-00855 CAAGK120000855 CAAG17-4130 AG17-4000 CAAG17-4025 CAAG17-4004 CAAG17-4045 CAAG17-4100 AG17-4015 CAAG17-4005 AG17-4130 CAAG17-4040 AG17-4050 CAAG17-4021 CAAG17-4120 AG17-4010 CAAG17-4140 CAAG17-4020 AG17-4110 AG17-4011 AG17-4055 AG17-4005 CAAG17-4015 AG17-4003 AG17-4025 AG17-4004 CA17-4001 CA17-4000 CA17-4003
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Dissolution Apparatus
Made of either color-coded UHMWPE or PVDF, the full flow filter prevents clogging.
  • Available in ultrahigh molecular weight polyethylene (UHMWPE) or polyvinylidene fluoride (PVDF)
  • Full flow filter offers increased surface area to prevent clogging
  • Low protein-binding PVDF filter material available
  • Usable with ¹/₈" diameter cannulas
  • Available in packs of 100 or 1000
  • Full flow filters are color coded for easy identification
  • Certificates of analysis can be provided on request
  • Standard conventional filter tips are made of polyethylene fluoride and can also be used with ¹/₈" diameter cannulas
Made of either color-coded UHMWPE or PVDF, the full flow filter offers increased surface area to optimize filter life and prevent clogging.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty