Would you like to log in again?
Splitless Liners for GC
Supplier: Agilent
CrossLab
5190-2270 5062-3587 210-3003-5 392611924 5183-4695 5183-4705 5181-3316 5190-2272 5181-3316I 5183-4706 18740-80220 5183-4694 5183-4694 5181-3316I 18740-80220 210-3003 5183-4703 5190-2271 210-3003-5 5183-4707 5181-8818 5183-4708 5181-3316 5183-4696 210-3003 5181-8818 5183-4703 5183-4705 5190-2271 392611924 5181-3315 5062-3587 5190-2270 5183-4708 5183-4693 5183-4704 5183-4696 5190-2272 5183-4707 5183-4693 5183-4704 5183-4706 5183-4695 5181-3315
AG18740-80220EA 46.78 USD InStock
AG18740-80220 CA76514-902 AG5183-4696 AG5183-4695 AG5183-4694 AG210-3003-5 AG5183-4693 AG5181-3316I CA76514-740 CA5183-4696 CA5183-4695 CA5183-4694 AG392611924 CA210-3003-5 CA5190-2271 AG5181-3315 CA76508-058 AG5181-3316 CA5183-4705 CA5183-4704 AG210-3003 CA210-3003 CA5183-4703 CAAG5190-2272 CA76514-738 CA5181-3316I CAAG5183-4706 CAAG392611924 CAAG5183-4707 CAAG5183-4708 CA76514-752 AG5183-4708 AG5183-4707 AG5183-4706 AG5183-4705 CA18740-80220 CAAG5190-2270 AG5062-3587 AG5181-8818 AG5190-2270 AG5183-4704 AG5183-4703 AG5190-2272 AG5190-2271
Splitless Liners for GC
Chromatography Supplies Chromatography Inlet Liners
We offer a complete line of liners for your Agilent GC splitless inlet, with exclusive deactivation.
  • Feature Agilent’s exclusive deactivation
  • Designed and engineered to meet strict specifications for dimensional accuracy and inertness
  • Outer diameter of splitless liners is ground and polished to exacting dimensional tolerances, ensuring a perfect fit into our inlets and optimal splitless injection
  • We recommend trying a liner without glass wool first
  • Use glass wool as a second choice
Choose a liner without glass wool if your sample contains nonvolatiles or analytes within a wide boiling range. For unknown samples, use a liner without glass wool to avoid loss of active or labile compounds.
Agilent offers a complete portfolio of liners for your Agilent GC splitless inlet. Our exclusive deactivation is important because of the longer sample/liner contact in splitless mode.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty