Would you like to log in again?
Graphite Furnace AAS Operating Supplies Kits, Agilent Technologies
Supplier: Agilent Technologies
190068000 190068000
AG190068000KT 9644 USD InStock
AG190068000 CAAG190068000
Graphite Furnace AAS Operating Supplies Kits, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Typical supplies for Agilent GTA 120 Deuterium and Zeeman graphite furnace AA Spectrometer systems.
  • Maintain accurate analysis
  • Save as much as 15% over the cost of purchasing supplies individually
  • Keep your instruments running for up to 12 months (depending on use) without worrying about reordering supplies
Agilent's spares kit provides the typical operating supplies required for moderate use of the GTA 120 deuterium and Zeeman graphite furnace Atomic Absorption systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty