Would you like to log in again?
J&W Ultra-1 Columns, Agilent Technologies
Supplier: Agilent
J&W
19091A-102E 19091A-012 19091A-002 19091A-101-INT 19091A-105 19091A-108 19091A-102 19091A-115 19091A-002 19091A-101 19091A-101 19091A-012 19091A-102E 19091A-105 19091A-115 19091A-101-INT 19091A-108 19091A-008 19091A-112 19091A-008 19091A-102 19091A-005 19091A-005 19091A-112
AG19091A-008EA 722.65 USD InStock
AG19091A-008 AG19091A-108 AG19091A-105 CAAG19091A-105 CAAG19091A-115 CAAG19091A-005 CAAG19091A-002 CAAG19091A-112 CAAG19091A-102 CAAG19091A-012 CAAG19091A101I CA19091A-108 CAAG19091A-008 AG19091A-102E AG19091A-115 CA19091A-101 AG19091A101INT AG19091A-005 AG19091A-002 AG19091A-101 AG19091A-112 AG19091A-102 AG19091A-012 CAAG19091A102E
J&W Ultra-1 Columns, Agilent Technologies
Chromatography Columns
A nonpolar column with tighter QC specifications that delivers excellent accuracy in a range of chromatographic applications.
  • 100% Dimethylpolysiloxane
  • Nonpolar
  • Equivalent to HP-1, with tighter specifications for retention index and capacity factors
  • Bonded, crosslinked, and solvent rinsable
  • Intended for use with the Sadtler Retention Index Library
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W Ultra 1 is a 100% dimethylpolysiloxane, nonpolar column that resembles HP-1 but has tighter specifications for retention index and capacity factors. The column has an operating temperature range of –60 - 325/350 °C and is intended for use with the Sadtler Retention Index Library.
These precision-engineered columns are bonded, crosslinked, and solvent rinsable, and are tested to some of the strictest industry QC standards, ensuring confidence in the accuracy and integrity of analytical results.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty