Would you like to log in again?
J&W Ultra-2 Columns, Agilent Technologies
Supplier: Agilent
J&W
19091B-105 19091B-101 19091B-012 19091B-102 19091B-102E 19091B-115 19091B-102E 19091B-012E 19091B-112 19091B-015 19091B-115 19091B-105 19091B-012 19091B-101 19091B-112 19091B-015 19091B-002 19091B-002 19091B-012E 19091B-102
AG19091B-012EA 1002.65 USD InStock
CA19091B-115 AG19091B-012 AG19091B-101 AG19091B-112 AG19091B-002 AG19091B-102 AG19091B-015 CAAG19091B012E CA19091B-102 CA19091B-101 CA19091B-112 AG19091B-102E AG19091B-115 AG19091B-105 CAAG19091B-015 AG19091B-012E CAAG19091B-105 CAAG19091B-012 CA19091B-102E CAAG19091B-002
J&W Ultra-2 Columns, Agilent Technologies
Chromatography Columns
Similar to HP-5 but with tighter specifications for retention index and capacity factors, for use with the Sadtler Retention Index Library.
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W Ultra 2 is a (5%-phenyl)-methylpolysiloxane, nonpolar column that resembles HP-5, but with tighter specifications for retention index and capacity factors. It has an operating temperature range of –60 to 325/350 °C. These precision-engineered columns are bonded, crosslinked, and solvent rinsable, and are tested to some of the strictest industry QC standards.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty