Would you like to log in again?
J&W HP-FFAP Columns, Agilent Technologies
Supplier: Agilent
J&W
19095F-120 19091F-112 19095F-123 19091F-433 19091F-115 19091F-102 19091F-105 19091F-112E 19091F-102 19095F-123E 19091F-115E 19091F-433 19091F-102E 19095F-123 19091F-105 19091F-112 19091F-413 19091F-115 19095F-123E 19091F-112E 19091F-102E 19095F-121 19095F-121 19091F-413 19091F-115E 19095F-120
AG19091F-115EEA 1445.29 USD InStock
AG19091F-115E AG19091F-115 CA19091F-102 CAAG19095F-121 AG19091F-413 AG19091F-112 AG19091F-433 CA19095F-123 CAAG19091F115E CA19095F-120 AG19095F-120 AG19095F-121 AG19095F-123 CAAG19091F-115 AG19091F-112E AG19091F-102E CA19091F-433 AG19095F-123E AG19091F-105 CA19091F-112 AG19091F-102 CA19091F-413 CAAG19091F102E CAAG19091F112E CAAG19095F123E CAAG19091F-105
J&W HP-FFAP Columns, Agilent Technologies
Chromatography Columns
HP-FFAP is a high-polarity phase designed for the analysis of volatile fatty acids and phenols.
  • Nitroterephthalic acid modified polyethylene glycol
  • Designed for the analysis of volatile fatty acids and phenols
  • Replaces OV-351
  • Bonded and cross-linked, solvent rinsable
  • Close equivalent to USP Phase G35
  • Similar Phases: Stabilwax-DA, Nukol, 007-FFAP, BP21, AT-1000, OV-351
Agilent J&W HP-FFAP is designed primarily for the analysis of organic acids, free fatty acids, or samples that require quantitation of acidic impurities. The FFAP stationary phase is modified with acid to provide a very inert column that can accommodate the demanding analysis of acids dissolved in water.;Free fatty acids up to C24 can be analyzed by HP-FFAP without costly and time-consuming derivatization. HP-FFAP does not require priming (several injections of acids), so accurate results can be obtained quickly.
Each Agilent J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results. For details about the QC test conditions including the test probe mixture and the resulting chromatogram, please refer to the Agilent J&W GC Column Performance Summary shipped with each GC column.
Note: We do not recommend the use of water or methanol to rinse HP-FFAP GC columns.
All columns are with 7" cage.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty