Would you like to log in again?
HP-5 Columns, Agilent Technologies
Supplier: Agilent
J&W
19091J-413E 19091J-215 19091J-112 19091J-233 19091J-577 19095J-621 19095J-123 19091J-205 19091J-102 19091J-416 19091J-436 19095J-323 19091J-215 19091J-231 19091J-413-INT 19095J-023 19091J-212 19091J-012E 19091J-433E 19091J-216 19091J-205 19091J-113 19091J-112E 19091J-101 19091J-413E 19091J-012 19091J-433E 19091J-216 19095J-321 19095J-323 19091J-433 19091J-433 19091J-236 19091J-202 19091J-216E 19091J-577LTM 19091J-216E 19091J-577 19095J-023 19091J-102 19091J-115 19091J-413 19091J-416 19091J-333 19091J-411E 19091J-213 19091J-413-INT 19095J-121 19091J-236 19091J-233 19091J-113 19091J-213 19095J-621 19091J-012 19091J-577LTM 19091J-215E 19091J-231 19091J-005 19091J-012E 19095J-123 19091J-411E 19091J-215E 19091J-413 19091J-101 19091J-112E 19091J-333 19091J-411 19091J-202 19091J-431 19095J-623 19091J-212 19091J-411 19091J-005 19095J-321 19091J-115 19091J-436 19091J-112 19095J-121 19095J-623 19091J-431
AG19091J-413EEA 869.85 USD InStock
AG19091J-413E AG19091J-233 AG19091J-431 AG19091J-112 AG19091J-433E AG19091J-433 AG19091J-113 AG19091J-231 CA19095J-321 AG19091J-215E CA19095J-121 CAAG19091J413I AG19091J-115 CA19091J-215 AG19091J-236 AG19091J-436 CA19091J-012 CAAG19091J-113 CAAG19091J-236 CAAG19091J-115 CAAG19091J-231 CAAG19091J-112 CAAG19091J-233 AG19095J-023 CAAG19091J216E AG19095J-621 CAAG19091J-436 AG19095J-623 AG19091J-216E AG19091J-202 CAAG19091J012E CAAG19095J-323 CAAG19091J577L AG19091J-005 AG19091J-205 CAAG19091J-102 CA19091J-101 CAAG19091J433E CAAG19095J-123 CAAG19091J215E CA19091J-413E AG19091J-012 CA19095J-623 AG19091J-411E AG19091J-213 AG19091J-411 AG19091J-212 AG19091J-333 CAAG19091J112E AG19095J-121 AG19091J-416 AG19091J413INT AG19091J-215 AG19091J-413 AG19091J-577 AG19091J-216 CAAG19091J-213 CAAG19091J-411 CA76514-570 CAAG19091J-333 CAAG19091J-212 CA19091J-431 CA19091J-433 CAAG19091J-577 AG19095J-123 AG19095J-321 AG19091J-112E AG19095J-323 CAAG19091J-216 CAAG19091J-416 AG19091J-101 AG19091J-102 CAAG19095J-621 AG19091J577LTM CAAG19091J-005 CA19091J-202 CAAG19095J-023 CAAG19091J411E AG19091J-012E CAAG19091J-205
HP-5 Columns, Agilent Technologies
Chromatography Columns
An excellent nonpolar, general-purpose 5% phenyl GC column with high temperature limits for a wide range of applications.
  • (5%-Phenyl)-methylpolysiloxane
  • An excellent nonpolar, general-purpose column for a wide range of applications
  • High temperature limit
  • Bonded, crosslinked, and solvent rinsible
  • Equivalent to USP phase G27
  • Wide range of column dimensions available
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W HP-5 is a precision-engineered (5%-phenyl)-methylpolysiloxane nonpolar column that delivers superb performance in a wide range of applications. This general-purpose column is bonded, crosslinked, and solvent rinsible and is equivalent to USP phase G27.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty