Would you like to log in again?
J&W HP-INNOWax Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
19091N-577IE 19091N-033I 19091N-131I 19091N-116I 19095N-123IE 19091N-030I 19095N-126I 19091N-216I 19091N-116I 19091N-236I 19091N-013I 19091N-133I 19095N-123IE 19091N-102I 19091N-231I 19091N-133I-INT 19091N-133IE 19091N-113I 19091N-136IE 19091N-131I 19091N-236I 19091N-205I 19091N-213I 19091N-136IE 19091N-205I 19091N-213I-INT 19091N-577I 19091N-216IE 19091N-133I-INT 19091N-133IE 19091N-033I 19091N-030I 19091N-105I 19091N-216IE 19091N-216I 19091N-233IE 19091N-233IE 19091N-213I 19095N-123I 19091N-013I 19091N-136I 19091N-111I 19091N-111I 19091N-105I 19091N-233I 19091N-113IE 19091N-133I 19091N-577I 19091N-577IE 19091N-233I 19095N-123I 19095N-121I 19091N-136I 19091N-213I-INT 19095N-126I 19091N-231I 19091N-113IE 19091N-102I 19091N-113I 19095N-121I
AG19095N-121IEA 495 USD InStock
AG19095N-121I AG19091N-233IE CA19091N-133I CAAG19091N205I CAAG19091N233E CAAG19091N213I CAAG19091N233I AG19091N-216IE CAAG19091N213N AG19091N-577I AG19091N-233I AG19091N-213I AG19091N-136I CAAG19095N126I CAAG19091N030I AG19091N-116I CAAG19091N131I AG19091N-205I CAAG19091N111I AG19091N-133IE CAAG19091N216E CAAG19091N216I CAAG19091N236I AG19091N-133I AG19091N-113I AG19091N-105I CAAG19095N121I CAAG19091N136I CAAG19091N116I CAAG19091N136E AG19091N213INT AG19095N-123I CAAG19091N231I AG19091N-231I AG19091N-013I AG19091N-033I CAAG19091N133N AG19091N-102I CAAG19091N133I CAAG19091N113I CAAG19091N105I AG19091N-577IE CAAG19091N113E AG19091N-136IE AG19091N-113IE CAAG19091N577E AG19095N-126I CAAG19091N577I AG19091N-030I AG19091N133INT AG19091N-131I AG19091N-111I CAAG19095N123E AG19091N-236I CAAG19091N033I AG19091N-216I CAAG19095N123I CAAG19091N013I CAAG19091N102I AG19095N-123IE
J&W HP-INNOWax Columns, Agilent Technologies
Chromatography Columns
A polyethylene glycol (PEG) column with a high upper temperature limit, ideal for food, flavor, and fragrance applications.
  • High polarity
  • Highest upper temperature limits of bonded PEG phases
  • Sharp and symmetrical peak shape performance for active polar compounds
  • Excellent column-to-column inertness reproducibility
  • Bonded, crosslinked, and solvent rinsable
  • Close equivalent to USP Phase G16
  • QC tested with highly demanding test probes
  • Performance Summary shipped with each column
HP-INNOWax is a polyethylene glycol (PEG) stationary phase that features high polarity and high upper temperature limits. It is a close equivalent to USP phase G16 and is targeted at food, flavor, and fragrance applications. Agilent J&W HP-INNOWax GC columns utilize the same innovative inert processes employed in the manufacture of Agilent J&W DB-WAX Ultra Inert columns. These columns have an extended inertness lifetime that withstands repeated temperature cycling to the upper temperature limits of the column, while selectivity, theoretical plates, and retention indices remain unchanged.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty