Would you like to log in again?
J&W HP-PLOT Al2O3 KCl Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
19091P-K15 19091P-K33 19095P-K25 19095P-K23 19095P-K23PT 19095P-K23 19091P-K33 19095P-K25 19095P-K23PT 19091P-K15
AG19091P-K15EA 1459 USD InStock
AG19091P-K15 CAAG19095P-K23 CAAG19091P-K33 CAAG19091P-K15 CAAG19095P-K25 AG19095P-K23 AG19091P-K33 AG19095P-K23PT AG19095P-K25 CAAG19095PK23P
J&W HP-PLOT Al2O3 KCl Columns, Agilent Technologies
Chromatography Columns
The column of choice for light hydrocarbons, and for the quantification of dienes.
  • Least polar alumina phase
  • Aluminum oxide deactivated with KCl
  • Low retention of olefins relative to comparable paraffin
  • Recommended phase for many ASTM methods
  • Preferred KCl deactivated alumina
  • Similar phases: Rt-Alumina PLOT, Alumina PLOT, Al₂O₃/KCl, AB-PLOT Al₂O₃/KCl, AT-Alumina
Agilent J&W HP-PLOT Al₂O₃ KCl is the least polar alumina phase and is deactivated with sodium chloride. It is the standard column choice for light hydrocarbon analysis, particularly C1 to C8 hydrocarbon isomers. Excellent for the quantification of dienes, especially propadiene and butadiene from ethylene and propylene streams.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty