Would you like to log in again?
HP-5ms Ultra Inert Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
19091S-133UI 19091S-433UI G3903-61005 G3903-61005 19091S-413UI-INT 19091S-436UI 19091S-413UI 19091S-133UI 19091S-436UI 19091S-413UI 19091S-413UI-INT 19091S-233UI 19091S-233UI 19091S-430UI-INT 19091S-431UI 19091S-431UI 19091S-433UI 19091S-430UI-INT 19091S-433UIE
AG19091S-431UIEA 503 USD InStock
AG19091S-431UI AG1909S430UINT AG19091S-413UI AG19091S-436UI AG19091S-233UI AG19091S-433UI CAAG19091S413N CA19091S-433UI CAAGG390361005 CAAG19091S413U AGG3903-61005 CA19091S-436UI CA19091S-431UI AG19091S-133UI AG1909S413UINT CAAG19091S233U CAAG19091S430U CAAG19091S133U CAAG19091S433U
HP-5ms Ultra Inert Columns, Agilent Technologies
Chromatography Columns
A nonpolar (5%-phenyl)-methylpolysiloxane, low-bleed GC column for trace-level analysis of acidic and basic compounds.
  • Excellent for trace-level analysis of acids and bases
  • (5%-Phenyl)-methylpolysiloxane has identical selectivity to HP-5MS
  • Specifically tested for inertness for active compounds, including acidic and basic compounds
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Nonpolar with very low bleed characteristics, ideal for GC/MS
  • Bonded, crosslinked, and solvent rinsible
  • Equivalent to USP phase G27
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Rtx-5ms, Rxi-5ms, Rxi-5Sil MS, PTE-5, BPX-5, AT-5ms, ZB-5ms, ZB-5ms Plus, SLB-5ms, Equity-7
Agilent J&W HP-5ms Ultra Inert is a (5%-phenyl)-methylpolysiloxane specifically tested for inertness in the analysis of active compounds, including acidic and basic compounds, to meet the growing need for lower detection levels. With very low bleed characteristics, this bonded and crosslinked column is solvent rinsible and ideal for GC/MS. It is equivalent to USP phase G27 and features improved signal-to-noise ratio for better sensitivity and mass spectral integrity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty