Would you like to log in again?
J&W Ultra Inert HP-1ms Columns, Agilent Technologies
Supplier: Agilent
J&W
19091S-733UI 19091S-911UI 19091S-933UI 19091S-612UI 19091S-933UI-INT 19091S-713UI 19091S-913UI-INT 19091S-933UI-INT 19091S-677UI 19091S-713UI 19091S-633UI 19091S-931UI 19091S-633UI 19091S-911UI-INT 19091S-911UI-INT 19091S-913UI 19091S-677UI 19091S-911UI 19091S-733UI 19091S-933UI 19091S-913UI 19091S-612UI 19091S-913UI-INT
AG19091S-933UIEA 845.15 USD InStock
AG19091S-933UI AG1909S933UINT AG19091S-911UI AG19091S-913UI AG19091S-713UI AG1909S911UINT AG19091S-677UI AG19091S-733UI CAAG19091S913I CAAG19091S933I AG19091S-931UI CAAG19091S911I AG1909S913UINT CAAG19091S913U AG19091S-633UI AG19091S-612UI CAAG19091S911U CAAG19091S933U CAAG19091S713U CAAG19091S612U CAAG19091S733U CAAG19091S633U CAAG19091S677U
J&W Ultra Inert HP-1ms Columns, Agilent Technologies
Chromatography Columns
Nonpolar PDMS with outstanding inertness that delivers excellent peak shapes for challenging active analytes.
  • Individually tested with Ultra Inert 1 ms test probe mixture
  • Consistent column inertness performance
  • Exceptionally low column bleed
  • Great peak shapes for challenging active analytes
  • Excellent signal-to-noise ratios
  • Minimum compound adsorption or degradation
  • Supports 0.18 mm id column configuration for higher sample throughput
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W HP-1ms Ultra Inert features exceptionally low column bleed and is designed to deliver consistent column inertness performance. They are individually tested with an Ultra Inert 1ms test probe mixture according to the strictest industry QC standards to ensure excellent signal-to-noise ratios and high-quality peak shapes for challenging active analytes. These columns deliver high levels of accuracy and results integrity in a range of applications, with minimal compound adsorption or degradation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty