Would you like to log in again?
HP-5ms Columns, Agilent Technologies
Supplier: Agilent
J&W
19091S-436 19091S-577UI-INT 19091S-113 19091S-416 19091S-333 19091S-413 19091S-105 19091S-231 19091S-577UI 19091S-577UI 19091S-331 19091S-577UI-INT 19091S-333 19091S-111LTM 19091S-233 19091S-102 19091S-436 19091S-101 19091S-336 19091S-102 19091S-577 19091S-213 19091S-112 19091S-336 19091S-101 19091S-133 19091S-213 19091S-413 19091S-233UI-INT 19091S-433-INT 19091S-433 19091S-112 19091S-433-INT 19091S-331 19091S-133 19091S-111LTM 19091S-416 19091S-431 19091S-113 19091S-431 19091S-433I 19091S-433E 19091S-139 19091S-139 19091S-577 19091S-105 19091S-433 19091S-233 19091S-233UI-INT 19091S-231
AG19091S-113EA 872.94 USD InStock
CA19091S-413 CA19091S-139 CAAG19091S-105 CAAG19091S433I AG19091S-113 AG19091S-233 AG19091S-112 AG19091S-431 AG19091S-231 AG19091S-105 CAAG19091S433N AG19091S111LTM AG19091S-577UI CAAG19091S-112 CAAG19091S-233 AG1909S577UINT CAAG19091S-113 AG1909S233UINT CAAG19091S-231 AG19091S433INT CAAG19091S-336 CAAG19091S-416 CA19091S-433E CA19091S-101 CA19091S-102 CA76514-574 CAAG19091S577U AG19091S-436 AG19091S-433 CAAG19091S577N CA19091S-436 CAAG19091S233I AG19091S-333 CA19091S-431 AG19091S-331 AG19091S-133 CAAG19091S-333 CAAG19091S-213 CAAG19091S-577 CAAG19091S-133 CAAG19091S-331 AG19091S-102 AG19091S-101 AG19091S-139 AG19091S-413 AG19091S-336 AG19091S-577 AG19091S-213 CAAG19091S111L AG19091S-416
HP-5ms Columns, Agilent Technologies
Chromatography Columns
Nonpolar, low-bleed columns with excellent inertness for active compounds, including acidic and basic compounds.
  • (5%-Phenyl)-methylpolysiloxane
  • Identical selectivity to HP-5
  • Nonpolar with very low bleed characteristics, ideal for GC/MS
  • Excellent inertness for active compounds, including acidic and basic compounds
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Bonded, crosslinked, and solvent rinsable
  • Equivalent to USP phase G27
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Rtx-5ms, Rxi-5ms, Rxi-5Sil MS, PTE-5, BPX-5, AT-5ms, ZB-5ms, ZB-5ms Plus, SLB-5ms, Equity-5
Agilent J&W HP-5ms is a (5%-phenyl)-methylpolysiloxane phase with very low bleed characteristics that is ideal for GC/MS. The column is bonded, crosslinked, and solvent rinsible, and has excellent inertness for active compounds, including acidic and basic compounds, with improved signal-to-noise ratio for better sensitivity and mass spectral integrity.
Equivalent to USP phase G27, it delivers excellent performance across a range of applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty