Would you like to log in again?
GC Columns, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
392372031 19095V-420 19095V-420 392372031
AG392372031EA 416 USD InStock
AG392372031 AG19095V-420 CA19095V-420 CAAG392372031
GC Columns, Agilent Technologies
Chromatography Columns
Agilent offers GC Columns for your special application needs.
  • Columns deliver excellent chromatographic performance
  • Wide range of particle sizes for flexible analysis
  • Exceptional stability for long column lifetime
Agilent J&W GC columns help lab analysts maintain the highest standards of performance. The columns deliver industry-leading technology with the highest inertness, lowest bleed levels, and tightest column-to-column reproducibility specifications.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty