Would you like to log in again?
Cool On-Column (COC) Inlet for GC, Agilent Technologies
Supplier: Agilent
9301-0658 19320-80625 19245-20525 19245-20510 19320-80625 19245-60760 19245-20515 19245-40050 5080-8853 19091-63000 G1545-80530 19245-80521 19091-63000 5080-8853 19245-20525 19245-80521 G1545-80530 19245-80521 19245-20510 19245-20780 19245-20515 19245-20580 19245-20780 G1545-80530 19245-20580 0100-1389 19245-60760 19245-40050 9301-0658
AG19245-20525EA 134.12 USD InStock
AG19245-20525 CAAG1924520510 AG19245-60760 AG19245-40050 AG19091-63000 CA76508-084 CAAG9301-0658 CAAG1932080625 AG9301-0658 CA19245-20515 AG19245-20580 AG19245-20780 CA19245-80521 AGG1545-80530 AG5080-8853 101479-642 101481-170 AG19245-20515 CA76514-578 CAAG1924540050 AG19320-80625 AG19245-80521 CA19245-20525 CA76508-026 CAAG1909163000 102945-150 CA19245-20780 CAAGG154580530 AG19245-20510
Cool On-Column (COC) Inlet for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Cool on-column inlets afford high precision injection to eliminate sample discrimination, sample alteration.
  • Elimination of sample discrimination
  • Elimination of sample alteration
  • Solvent focusing of early eluting solutes
  • High analytical precision
Cool on-column injection is superior in many ways to other sample introduction techniques by offering high precision injection to eliminate sample discrimination and sample alteration. Agilent provides all the necessary supplies to set-up and maintain your COC injection port.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty