Would you like to log in again?
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
G8020-60808 2010095900 G8020-60807 G8020-60808 G8020-60809 2010095900 2010101700 G8020-60805 2410067590 G8020-60809 G8020-60806 2010095400 2010077500 2010096000 2410067590 G8020-60805 2010096000 2010101700 2010095600 G8020-60807 2010077500 2010095400 2010095600 G8020-60806
AG2410067590EA 182 USD InStock
CAAG2010095900 CAAG2010095600 CAAG2010101700 AG2410067590 CAAG2010095400 AG2010077500 CAAG2010096000 AGG8020-60808 AGG8020-60809 AG2010095900 CAG8020-60806 AGG8020-60805 AGG8020-60806 CAAG2010077500 AG2010095600 AGG8020-60807 AG2010095400 CAAG2410067590 CAAGG802060809 AG2010096000 CAAGG802060808 CAAGG802060807 AG2010101700 CAAGG802060805
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
ICP-OES quartz and ceramic injectors for demountable torches.
  • A variety of injector sizes and materials to handle difficult and diverse application demands
  • Removable injectors provide quick change-over without the need for diagrams or multiple o-rings and seals
  • No optimization required with the Agilent fully demountable torch providing consistent results before and after injector replacement regardless of number of analysts
  • Greater productivity Tool free installation simplifies injector maintenence
Agilent ICP-OES injectors for fully demountable torches are guaranteed to fit with your Agilent instrument. ICP-OES injectors are available in quartz and alumina with a variety of inner diameters (0.8 mm, 1.4 mm, 1.8 mm, 2.4 mm) to meet your application needs. Analysis of organic solvents require smaller inner diameter injector such as the 0.8 mm and 1.4 mm injector. Aqueous analysis uses a 1.8 mm quartz injector while high total dissolved solids equires the 2.4 mm injector. When analysing HF an inert material is required such as the ceramic alumina injector.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty