Would you like to log in again?
CrossLab Nebulizers for ICP-OES, Agilent Technologies
Supplier: Agilent
CrossLab
2010096400 G8010-60270 2010097600 CP914506 2010106800 2010126000 2010097600 G8010-60255 G8010-60255 2010097700 2010097700 G8010-60346 2010096400 G8010-60346 G8010-60293 G8010-60347 G8010-60270 G8010-60347 2010106800 2010126000 CP914506 G8010-60293
AGCP914506EA 1548.19 USD InStock
AGCP914506 CA2010096400 CAAGG801060270 AGG8010-60270 CAAG2010106800 AGG8010-60293 AG2010126000 CAAGG801060346 CAAGCP914506 AG2010106800 CAG8010-60293 AG2010097700 CAAGG801060347 AG2010097600 AG2010096400 CAAG2010097600 AGG8010-60255 CAAG2010097700 CAG8010-60255 AGG8010-60346 CAAG2010126000 AGG8010-60347
CrossLab Nebulizers for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Glass concentric, Seaspray, Miramist, and inert One Neb 2 nebulizer offering superior performance.
  • Manufactured to fine tolerances for reproducible and stable sample uptake
  • Tight specifications deliver fine droplets for best signal intensity and optimum signal to noise
  • Fewer repeats means less downtime and improved productivity
The universal OneNeb Series 2 nebulizer provides improved sensitivity and presision, greater tolerance to disolved solids, and is inert so it is suitable for use with aggressive acids such as hydrofluoric acid (HF) and most common organic solvents. Agilent nebulizers and accessories have tight specifications for delivering fine droplets for the best signal intensity and optimum signal-to-noise. The Seaspray glass concentric nebulizer comes standard on the ICP-OES instrment. The unique OneNeb Series 2 nebulizer provides improved sensitivity and provides greater tolerance to dissolved solids. The OneNeb Series 2 nebulizer is a universal, high-efficiency nebulizer offering superior performance with ICP-OES instruments. Based on flow-blurring nebulization technology, the nebulizer provides improved sensitivity, precision, and greater tolerance to dissolved solids. The inert construction makes the OneNeb capable to handle most ICP-OES sample matrices including strong acids.
The OneNeb Series 2 nebulizer is a universal, high-efficiency nebulizer offering superior performance with ICP-OES instruments. Based on flow-blurring nebulization technology, the nebulizer provides improved sensitivity, precision, and greater tolerance to dissolved solids. The inert construction makes the OneNeb capable to handle most ICP-OES sample matrices including strong acids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty