Would you like to log in again?
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Supplier: Agilent
CrossLab
27-0103 33-9025 27-0106 27-8621 33-9022 27-0103 27-8036 27-0102 33-9009 33-9048 27-8600 27-3000 33-9041 33-9018 27-0105 27-0104 33-9048 27-0100 27-8020 33-9051 27-0102 27-3002 27-0108 33-9046 27-0107 27-0104 33-9007 27-8020 27-0101 33-9012 27-8005 27-8622 33-9022 27-0105 33-9018 33-9051 33-9007 33-9052 27-3000 27-0109 33-9052 33-9025 27-8622 33-9009 27-3002 27-8620 27-3001 27-8005 33-9046 33-9029 33-9029 27-0109 27-0108 27-8620 27-0101 27-0100 27-0106 27-3001 33-9012 27-8600 27-0107 27-8036 27-8621
AG27-3000KT 610.57 USD InStock
CAAG27-8020 AG27-3000 AG33-9007 CAAG33-9018 AG33-9009 CAAG33-9012 AG33-9046 AG33-9048 AG33-9012 AG33-9052 AG33-9051 CAAG27-3000 CAAG27-3001 AG27-8600 CAAG27-3002 AG27-8005 AG33-9018 CAAG33-9029 CAAG33-9022 CAAG33-9025 AG33-9022 CAAG27-8600 CAAG27-8005 AG27-8036 AG33-9029 AG33-9025 AG27-0103 AG27-0104 AG27-0101 AG27-0102 AG27-0100 AG27-0109 AG27-8622 AG27-0107 AG27-0108 AG27-0105 AG27-0106 AG27-8020 CAAG33-9009 AG27-8620 CAAG27-8036 AG27-8621 CAAG33-9048 CAAG33-9007 CAAG33-9046 CAAG27-0101 CAAG33-9052 CAAG33-9051 CAAG27-0102 CAAG27-0100 AG33-9041 AG27-3002 AG27-3001 CAAG27-8622 CAAG27-0109 CAAG27-0107 CAAG27-8620 CAAG27-8621 CAAG27-0108 CAAG27-0105 CAAG27-0106 CAAG27-0103 CAAG27-0104
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Dissolution Apparatus
Agilent can provide many different dosage form holders for the Apparatus 7.
  • Accessories for the reciprocating holder apparatus 7
  • Simulates the biorelevant conditions for the skin, gastrointestinal tract and tissue required for transdermal, oral drugs and implants with biorelevant temperature agitation rates and retention times
  • Accommodates a variety of dosage form holders including reciprocating disks, angled disks, spring holders, stent holders, and pointed rods
  • Supports typical volume configurations of 5 to 300 ml, depending on instrument
Ideal for automatic dissolution testing of dosage forms that require a change of media, a smaller volume, or more vigorous agitation. The Agilent 400-DS dissolution apparatus 7 enables of small-volume drug-release testing of medical devices such as drug eluting stents (DES).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty