Would you like to log in again?
Dissolution Evaporation Covers, Agilent Technologies
Supplier: Agilent Technologies
12-6327 12-6328 12-6315 12-6315 12-6311 12-6330 27-1500 12-6328 12-6305 12-6300 12-6311 12-6310 12-6305 12-6330 27-1510 12-6327 12-6329 12-6300 12-6320 12-6329 12-6320 12-6310 27-1500 27-1510
AG12-6329EA 323.93 USD InStock
CAAG12-6328 AG12-6329 CAAG12-6305 CAAG12-6327 AG12-6327 CAAG12-6315 AG12-6305 AG12-6328 CAAG12-6329 CAAG12-6320 AG12-6310 AG12-6311 CAAG12-6330 AG12-6300 AG12-6330 AG12-6320 AG12-6315 CAAG12-6311 CAAG12-6300 CAAG12-6310 AG27-1500 CAAG27-1510 AG27-1510 CAAG27-1500
Dissolution Evaporation Covers, Agilent Technologies
Dissolution Apparatus
A variety of evaporation covers are available to fit your apparatus and vessels.
  • The evaporation covers automatically travel with the apparatus driveunit using either the alignment posts or DDM assemblies
  • The standard evaporation cover on the 708-DS is designed to allow less than 1% evaporative loss over a 24-hour period under normal operating conditions
  • Agilent also offers other low loss covers that can reduce evaporative loss even further
A variety of evaporation covers are available to fit your apparatus and vessels. Evaporation plugs are used to minimize evaporation by sealing unused ports in the evaporation covers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty