Would you like to log in again?
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: Agilent
CrossLab
27-2315 27-2000 27-2110 27-2007 27-2103 27-2205 27-2005 27-2105 27-2007 27-2115 27-2210 27-2103 27-2200 27-2200 27-2105 27-2015 27-2310 27-2010 27-2120 27-2215 27-2210 27-2315 27-2205 27-2010 27-2310 27-2100 27-2115 27-2215 27-2100 27-2110 27-2015 27-2005 27-2000 27-2120
AG27-2010PK 284.51 USD InStock
AG27-2010 AG27-2205 AG27-2007 CAAG27-2115 AG27-2105 CAAG27-2015 AG27-2005 AG27-2103 CAAG27-2310 CA27-2005 CAAG27-2210 CAAG27-2110 AG27-2200 AG27-2100 AG27-2000 CAAG27-2315 CAAG27-2215 AG27-2120 AG27-2315 CAAG27-2103 AG27-2215 AG27-2115 CAAG27-2200 AG27-2015 CAAG27-2100 CAAG27-2000 AG27-2310 AG27-2210 CAAG27-2120 AG27-2110 CAAG27-2007 CA27-2010 CAAG27-2205 CAAG27-2105
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Dissolution Apparatus
Several reciprocating vessel screens are available for the BIO-DIS reciprocating cylinder apparatus.
  • Reciprocating cylinder screens for BIO-DIS in stainless steel and polypropylene are available in different mesh sizes
Several reciprocating vessel screens are available for the Agilent BIO-DIS reciprocating cylinder apparatus, which is ideal for automatic dissolution testing of dosage forms that require different types of media.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty