Would you like to log in again?
CrossLab Atomic Absorption Tubing, Agilent Technologies
Supplier: Agilent
CrossLab
3710026800 3710026600 3710034000 3710026600 3710009200 3710026800 3710009200 3710034000
AG3710026800EA 254.67 USD InStock
CAAG3710034000 AG3710026800 AG3710009200 AG3710034000 CAAG3710009200 CAAG3710026600 AG3710026600 CAAG3710026800
CrossLab Atomic Absorption Tubing, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Drain tubing from the liquid trap to waste vessel for the Mark 7 & Mark VI flame atomization systems.
  • Regular inspection of the drain tubing used with the flame atomization system is recommended
  • The drain tubing should be replaced if the tubing has perished, or shows excessive wear or discoloration
  • The regular clear vinyl drain tubing is suitable for use with aqueous solutions, including acid digests
  • For compatibility with organic solvents, black rubber drain tubing should be fitted
Drain tubing is used with the current Agilent Mark 7 flame atomization system, and the earlier Agilent Mark VI flame atomization system, to take the liquid waste from the liquid outlet on the liquid trap to the drain or waste vessel. Regular inspection of the drain tubing is recommended, especially when completing routine maintenance on the flame atomization system. The drain tubing should be replaced if the tubing has perished, or shows excessive wear or discoloration. The regular 'clear' vinyl drain tubing is suitable for use with aqueous solutions, including acid digests. For compatibility with organic solvents, black rubber drain tubing should be fitted.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty