PVC, Solvaflex, Marprene and FPM peristaltic pump tubing for ICP-OES.
- Standard PVC suitable for all aqueous with medium concentrated acids or bases
- PVC Solvaflex for common organic solvents such as kerosene, xylene, white spirit, alcohols, medium-highly concentrated acids and bases
- Marprene tubing for ketone-based solvents such as MIBK and DIBK;FPM Fluoroelastomer Polymer tubing is suitable for gasoline, naphtha, toluene, xylene (aromatic hydrocarbons) and highly concentrated acids and bases
Agilent top-quality ICP-OES Peristaltic Pump Tubing allows you to find the appropriate product to handle any matrix. Agilent has tubing in the diameters and material required for your application: PVC, Solvaflex, FPM, or Marprene.
;Standard PVC works for all aqueous with medium concentrated acids or bases, PVC Solvaflex works for common organic solvents such as kerosene, xylene and white spirit. Marprene tubing is for ketone-based solvents such as MIBK and DIBK. FPM Fluoroelastomer Polymer tubing is for gasoline, naphtha, toluene, xylene (aromatic hydrocarbons) and highly concentrated acids and bases.