Would you like to log in again?
CrossLab Vacuum Pump Supplies for GC/MS, Agilent Technologies
Supplier: Agilent
CrossLab
392028900 3162-1056 1535-4970 3162-1056 393064601 G3170-80006 G1099-80039 1535-4970 G1099-80039 G3170-80006 392028900 393064601
AG3162-1056KT 339.23 USD InStock
CAAG393064601 AG3162-1056 CA3162-1056 AG392028900 AG1535-4970 AGG1099-80039 CAAGG317080006 AGG3170-80006 CAG1099-80039 AG393064601 CA1535-4970 CAAG392028900
CrossLab Vacuum Pump Supplies for GC/MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A range of vacuum pump supplies to ensure the accuracy and performance of your GC/MS system.
  • Oil mist and exhaust filters for vacuum and rotary vane pumps;Oil for turbo, diffusion, and foreline pumps
  • Further accessories including oil return lines, oil pans, vacuum gauge tubes
Agilent provides a range of vacuum pumps and supplies to ensure the highest levels of accuracy and performance of our GC/MS systems. These supplies include foreline and diffusion pump supplies such as pump oil, oil mist filter kits, and gauge tubes for measuring vacuum. Tubing and additional pump supplies are also available, and are manufactured to high levels of quality and consistency.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty