Would you like to log in again?
Agilent Prep HPLC Columns, Agilent Technologies
Supplier: Agilent
413910-502 820675-119 410910-102 443905-102 446905-902 449905-101 420212-902 413910-302 446905-901 449905-502 410910-501 410910-502 446905-301 449905-501 443905-101 449910-902 449905-301 440910-901 419910-502 440910-902 449905-901 884950-547 419910-302 413910-301 443910-902 413910-101 419910-301 443910-901 419910-501 446910-102 410910-301 446905-501 446905-102 880975-201 420212-902 446905-101 880952-206 440905-901 413910-501 440905-902 880952-202 449905-102 413910-102
101471-432EA 2161.61 USD InStock
101471-432 AG420212-902 101471-498 101471-454 101471-476 101471-496 101471-452 101471-474 101471-430 101471-458 101471-436 101471-478 101471-434 101471-456 101483-742 CA420212-902 101483-826 101471-480 101473-456 101473-454 101471-440 101483-306 101473-452 101471-482 101473-450 101471-466 101471-422 101471-488 101471-444 101471-442 101471-448 101471-502 101471-424 101471-446 101471-500 101471-940 101471-428 101473-448 101471-490 101483-734 101471-450 101471-472 101483-956
Agilent Prep HPLC Columns, Agilent Technologies
Chromatography Columns
Preparative LC Columns are designed for high loadability to purify miligram to gram quantities of products.
  • Easy scalability from 4.6 to 50 mm Int.Ø for rapid method development
  • High throughput 21.2 mm Int.Ø cartridges for fast purification
  • Exceptional column stability up to pH 10
Column dimensions are available for most preparative sample types ranging from high throughput samples generated in drug discovery to lab scale prep and up to some pilot scale sample preparation. Preparative sized columns are available in 21.2, 30 and 50 mm internal diameters with lengths ranging from 50 to 250 mm. Columns are available in 5 and 10 µm particle sizes with very high efficiency in every dimension.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty