Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CrossLab Peristaltic Pump Tubing for ICP-MS, Agilent Technologies

Supplier: Agilent Technologies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

5005-0025 G1820-65216 5005-0022 5005-0026 5005-0027 5042-4799 G1833-65571 5005-0022 R020031100 5005-0025 G1820-65216 G1820-65217 5005-0020 5005-0021 G1833-65571 G3280-67008 5005-0023 5042-4799 R020031100 5005-0020 5005-0026 G3280-67008 5005-0021 5005-0023 5005-0027 G1820-65217
AGG1820-65217PK 237 USD InStock
CAR020031100 AGG1820-65217 AG5042-4799 AGG1820-65216 AG5005-0027 AG5005-0026 AG5005-0025 CAG3280-67008 AG5005-0023 AG5005-0022 AGG1833-65571 CA76515-246 AG5005-0021 AG5005-0020 CAG1820-65217 CAAG5005-0026 CAAG5005-0025 CAAG5005-0027 CAAG5005-0021 CAAG5005-0023 CAAG5042-4799 AGR020031100 CAG1833-65571 CA5005-0020 AGG3280-67008 CA5005-0022
CrossLab Peristaltic Pump Tubing for ICP-MS, Agilent Technologies
Tubing Peristaltic Pump Tubing
Agilent offers a range of ICP-MS peristaltic pump tubing, in suitable materials for your application.

  • PVC tubing is the general purpose or standard tubing used for sample introduction and internal standard (ISTD) uptake
  • PVC tubing can be used for almost all aqueous reagents used in ICP-MS including HF
  • Lifetime of the PVC tubing is about one to two weeks when 1% HNO<sub>3</sub> is analyzed
  • Shorter lifetimes result when higher acid concentration are used for analyzes, or when the pressure on the peristaltic pump pressure bar is increased
  • Ismaprene tubing is good for almost all reagents and has about a 10 times longer lifetime compared to PVC
  • Fluran tubing is recommended for xylene analysis with yellow/blue tabs (1.52 mm Int.Ø)

However, it has many impurities so it is routinely recommended for the spray chamber drain to achieve efficient waste removal for a long time. The blue/orange PVC tube (0.25 mm Int.Ø) is used for online addition of internal standard solution. For high matrix samples to achieve 1:1 dilution online with ISTD we recommend black/black PVC (0.76 mm Int.Ø) tubing

Agilent provides a range of peristaltic pump tubing for ICP-MS, in suitable materials for your sample and matrix. This includes robust long-life drain tubing and high purity uptake tubing. Including materials such as PVC, SolvaFlex and Ismaprene to meet your application needs including aqueous, organic solvent or HF resistance.

The Agilent ICP-MS features an on-board peristaltic pump that controls the flow of solution into and waste out of the sample introduction system. Using a peristaltic pump to assist sample introduction reduces the variation in uptake flow rate due to variation in physical properties of the solutions being introduced.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty