Would you like to log in again?
CrossLab Solvent Inlet Tubing and Filters for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
280959-904 G1311-60006 79880-22711 5067-1551 5067-6189 01090-68703 01090-27609 5067-1553 01090-68702 5043-1790 280959-907 5022-2166 5022-2166 5067-6189 01090-27609 5043-1789 5067-5443 G4204-40000 5067-1562 5064-8273 5064-8273 5022-2185 G1311-60006 5067-1562 G7120-68070 G4204-40000 01090-68702 5065-4500 280959-904 0101-1001 280959-006 280959-907 01090-68703 5065-4500 UCA100X 5023-0271 79880-22711 5067-1551 5022-2165 RF0089T 280959-001 5067-1555 5022-2185 5067-5443 5023-0271 5067-1553 RF0089T 5067-1555 G7120-68070 5043-1790 5022-2165 5043-1789 UCA100X 0101-1001
AG5064-8273EA 297.3 USD InStock
CA5067-6189 AG5064-8273 AG0101-1001 CA76514-590 AGRF0089T CAAG0101-1001 AG5067-1555 AG5067-1553 CAAG5043-1789 AG5067-1551 CA01090-68702 CA01090-68703 AG5022-2185 CA5064-8273 CA5067-1562 AGG4204-40000 CAAGUCA100X CAAG5022-2165 CAAG5022-2166 CA5023-0271 CAAGG131160006 AGG1311-60006 AG280959-006 AG280959-001 CA280959-907 AG5067-1562 AG5043-1790 CA01090-27609 AG5065-4500 AG01090-27609 AGUCA100X AG01090-68703 AG01090-68702 CA79880-22711 CA5067-1551 CA5067-1553 CA5067-1555 AG5067-5443 CAAG5043-1790 CAAG5065-4500 CAAG5067-5443 AG5022-2165 AG5022-2166 AG5023-0271 AGG7120-68070 CA5022-2185 AG280959-907 AG280959-904 CAAGG712068070 AG5067-6189 CAAGRF0089T AG5043-1789 CAAGG420440000 AG79880-22711
CrossLab Solvent Inlet Tubing and Filters for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
High-quality glass and stainless steel solvent filters with different pore sizes for LC applications.
  • Solvent inlet filters represent the first barrier for retaining particles, precipitates, and microbes from mobile phases, buffers, and salt solutions
  • Filters are significant elements for preventing system blockage, pressure increases, and contamination
  • Agilent solvent filters have a defined, homogeneous pore size to effectively block particles above a certain size, while letting the mobile phase through without a significant pressure increase
Agilent provides a comprehensive range of high-quality glass and stainless steel solvent filters with different pore sizes, suitable for many different LC applications and equipment, to optimize HPLC results by removing any trace of particles from the solvent.
All solvent filters and frit adapters are manufactured to Agilent's high levels of quality and have been tested on Agilent instruments to ensure performance that will significantly increase your analytical, instrument, and laboratory efficiency.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty