Would you like to log in again?
CrossLab Sample Trays for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
G1364-84525 G1313-60004 G4226-60021 G1364-84524 G9321-60201 G1364-84516 5022-6531 G7129-60010 5023-2471 G7167-60020 G1313-27302 G2258-60111 G7129-68210 5023-2471 G1364-84523 G2258-60111 G9321-60201 G4226-60021 G7167-60020 G1313-27302 G7129-60110 G1364-84525 G7129-60110 5022-6531 G7129-60000 G1364-84516 G1364-84523 G7129-68210 G1313-60004 G1364-84524 G7129-60010 G7129-60000
AGG7167-60020EA 1226.72 USD InStock
CAAGG712960000 AGG7167-60020 AGG7129-60000 CAG1364-84525 CAAGG131360004 CAG1364-84524 CAG1364-84523 AGG1364-84524 AGG1364-84523 AGG2258-60111 AGG1364-84525 CAAGG422660021 AG5022-6531 CAAG5023-2471 CAAGG712960010 CAAGG712960110 AG5023-2471 AGG7129-60110 CAG1364-84516 AGG7129-60010 CAG7167-60020 CAAGG932160201 CAAGG225860111 AGG1313-60004 AGG1313-27302 AGG4226-60021 CA5022-6531 AGG1364-84516 AGG7129-68210 CAAGG131327302 CAAGG712968210 AGG9321-60201
CrossLab Sample Trays for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Designed to ensure that your autosampler can accommodate a variety of applications.
  • Agilent provides various trays with different hole sizes and different hole numbers to best suit all your applications
  • Available options include 100 position trays, thermostatted trays, 40 position trays, and 15 position trays for 6 ml vials
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
Agilent autosampler vial trays and drawers are designed to ensure that your autosampler setup is perfectly equipped to deal with a wide variety of applications and sample volumes.
Vial drawers are available with various sample position numbers, depending on the required capacity and volume of the vials used. All parts are manufactured to Agilent's highest levels of quality and have been tested in Agilent instruments to ensure they work perfectly with the rest of the system.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty