Would you like to log in again?
Manual Crimpers and Decappers, Agilent Technologies
Supplier: Agilent Technologies
8710-1643 5040-4669 5181-8827 5040-4667 5040-4669 5040-4668 5040-4671 8710-1643 5040-4667 5181-8827 5040-4668 5040-4671
AG5040-4667EA 292 USD InStock
CAAG5181-8827 AG5040-4667 AG5181-8827 CAAG8710-1643 AG5040-4668 AG5040-4669 CA5040-4671 CA5040-4667 CA5040-4669 AG5040-4671 AG8710-1643 CA5040-4668
Manual Crimpers and Decappers, Agilent Technologies
Crimpers, Cappers and Decappers
A selection of ergonomically designed manual vial crimpers, vial decappers, and related accessories.
  • Comfortable, lightweight, ergonomically designed handles fit smoothly in the hand
  • Top-mounted adjustment knob show directionality for tightening/loosening
  • Adjustment knob doubles as an indicator that the crimp (or decap) is complete
  • Crimpers are color-coded with blue knobs and labels, decappers with orange
  • Narrow jaws provide better vertical clearance over vials
  • Bottom handle motion allows for better control and enhanced stability of crimping jaw
  • Sturdy construction of rugged, fiber-reinforced resin with steel reinforcement in the handles
Agilent ergonomic manual vial crimpers and decappers remove the pain and discomfort of wrist strain with a lightweight, tailored design. Weighing 25 to 30% less than predecessors, the new design dramatically improves your experience by helping to eliminate sore, pinched hands. Extensively tested on both 11 mm crimp caps and 20 mm crimp caps, these vial crimpers are an economical alternative to electronic vial versions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty