Would you like to log in again?
CrossLab Fittings for HPLC, Agilent Technologies
Supplier: Agilent
8710-1217 RE6000282MC 5067-4738 5043-0259 UCP628 5067-6141 5067-1547 5067-5695 5042-6500 5065-4410 5067-4739 8710-1217 RF0032T 5067-1540 5005-0068 0100-1259 5065-9948 5067-1547 5067-1585 5023-0232 5062-8541 5062-8541 RF0031T 5067-4733 5062-2418 0100-1516 RE6000282 5065-9948 5065-4422 5022-6536 G1314-65062 RF0031T 5067-4733 5065-4423 5042-8957 5062-8518 5180-4114 5067-1585 5180-4114 0100-1631 5063-6591 UCP316 5067-6141 5043-0277 01080-83202 5065-4423 RF0100T 5062-8518 UCF130 0100-1259 5065-9967 5065-9950 5063-6593 5067-4739 5043-0277 5067-1557 UCP628 0100-1516 5065-4426 5005-0067 RE6000282 5022-6536 5005-0068 01080-83202 5042-6500 5067-1557 5065-9950 5067-1540 5065-4454 5023-0232 5067-1558 5065-4410 RE6000282MC 5065-4426 5042-8957 5043-0259 5180-4108 RF0100T 5063-6593 RF0030T 5067-5695 G1314-65062 UCP316 5065-4454 RF0030T 0100-1631 5067-4738 5062-2418 5180-4108 RF0032T RF0098T RF0098T 5065-4422 5067-1558 UCF130 5005-0067 5065-9967 5063-6591
AG5062-8541PK 118.28 USD InStock
CAAG5067-4739 AG5062-8541 CA5065-4422 AG5067-1540 AGG1314-65062 AGUCP628 CA5065-4426 CA76514-670 CAAG5067-1585 AG5067-1547 CAAGRF0030T AG5180-4114 CA0100-1631 CAAG8710-1217 AG5063-6593 CA5042-8957 AG5063-6591 CAAGRF0100T AGRF0030T AG5067-1558 AG5067-1557 CAAGRF0031T CAAG5065-9950 CA5062-8541 CA76514-494 CA5067-1557 CA5067-1558 CAAG5065-4423 CA0100-1259 CA5043-0277 CA76514-512 AG0100-1631 AG5065-4454 AG0100-1516 CAAG5005-0068 CAAG5005-0067 AG5067-6141 AG5067-5695 AGRE6000282 AG5043-0259 CAAG5067-1547 AG5067-1585 AG5067-4738 AG5067-4739 CA5067-6141 AG5042-8957 CA5067-5695 AGUCP316 CAAG5067-1540 CAAGRF0098T AG5067-4733 AG01080-83202 AGUCF130 AG5043-0277 CAAG5043-0259 CAAG5065-4410 AGRE6000282MC CAAG5023-0232 CAAGUCF130 AG5065-9967 AG0100-1259 AGRF0098T CAAG5065-9948 CAAGRE600282MC CAAGRE6000282 CAAGUCP628 AG5062-2418 AG5062-8518 AG5065-9950 AG5065-4423 CAAG5063-6593 AG5065-4422 CA76514-720 CAAGG131465062 AG5005-0068 AG5065-4426 AG5005-0067 CA5042-6500 AGRF0100T AGRF0031T CAAGRF0032T AG5022-6536 CA76514-718 AG8710-1217 CA5062-8518 AG5065-4410 CA5065-9967 CA76514-674 AG5023-0232 CA76508-050 AG5042-6500 CAUCP316 AG5180-4108 CA5022-6536 CA5067-4733 AG5065-9948 CA5067-4738 AGRF0032T
CrossLab Fittings for HPLC, Agilent Technologies
Fittings and Connectors
Fittings made of different materials to ensure reliable sealing and optimal performance in your LC.
  • Innovative InfinityLab Quick Connect and Quick Turn fittings providing finger-tight, reusable, and zero-dead-volume connections
  • A variety of fitting materials are available, including stainless steel, PEEK, and polyketone, for different pressure ranges and positions
  • Different dimensions covering Swagelok-type or metric M4/M3-type connection ports
  • An array of nuts, ferrules, various color identifiers, and other supplies are also available for all fitting types
Agilent has created a large variety of fittings made of different materials to ensure the ongoing security and performance of your LC equipment. Utilizing the correct fitting style and format, as specified by the manufacturer, is important for ensuring accuracy and robust performance in every aspect of your apparatus.;InfinityLab Quick Connect fittings offer truly finger-tight connections up to 1300 bar, for column connections as easy as pressing a lever. For harder-to-reach areas of the LC instrument, try InfinityLab Quick Turn fittings. You can find the right capillaries quickly and easily using our online selection tool.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty