Would you like to log in again?
Stay Safe Caps & Accessories for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
5043-1217 5043-1336 5043-1333 5043-1220 5043-1698 5043-1218 5043-1338 5043-1222 5043-1334 5043-1217 5043-1220 5043-1222 5043-1219 5043-1695 5043-1698 5043-1333 5043-1338 5043-1218 5043-1336 5043-1219 5043-1334 5043-1695
AG5043-1333KT 139.59 USD InStock
AG5043-1333 CAAG5043-1336 AG5043-1222 AG5043-1695 AG5043-1220 CAAG5043-1338 CAAG5043-1333 CAAG5043-1695 CAAG5043-1698 CAAG5043-1334 CAAG5043-1219 CA5043-1222 AG5043-1219 AG5043-1218 AG5043-1217 AG5043-1338 CAAG5043-1220 CA5043-1217 CA5043-1218 AG5043-1336 AG5043-1698 AG5043-1334
Stay Safe Caps & Accessories for HPLC, Agilent Technologies
Closures Safety Caps and Safety Waste Caps
Designed to stop evaporation of harmful vapors from solvent bottles.
  • Open or partially covered solvent bottles can lead to the evaporation of solvents and harmful solvent vapors
  • InfinityLab Stay Safe caps stop solvents from leaching into the air
  • Over time, solvent composition can change, affecting your chromatographic results
  • Storing solvents with airtight Stay Safe caps prevents this problem, ensuring the long-term consistency of your mobile phase for more reproducible results
InfinityLab Stay Safe caps stop evaporation of harmful solvent vapors from solvent bottles and stop solvent from leaching into the air. They have a GL 45 thread and are compatible with Agilent solvent bottles. InfinityLab Stay Safe caps include an easy-to-see time strip (time-based indicator) that tells you when the venting valve or the charcoal filter needs to be replaced.
Agilent InfinityLab products are designed to provide you the best efficiency in your liquid chromatography workflow, regardless of application area. When relying on Agilent InfinityLab instruments, columns, and supplies, be assured that every part works together seamlessly and supports your efforts to significantly increase your analytical, instrument, and laboratory efficiency in your everyday work.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty