Would you like to log in again?
CrossLab GC Fittings, Agilent Technologies
Supplier: Agilent
5180-4105 0100-0071 5180-4105 0100-0161 5180-4124 5180-4128 0100-0969 5180-4160 0100-0090 PL1310-0003 5180-4131 0100-0161 5180-4127 0100-0088 VLZT1C 5180-4161 G1544-20590 5180-4128 0100-0128 0100-0053 0100-0542 5180-4129 0100-0090 PL1310-0003 0100-0124 5180-4121 0100-0128 5061-3303 G1544-20590 5061-3303 0100-0053 5180-4121 5180-4127 0100-0088 0100-2144 5080-8751 0100-0969 5080-8751 0100-0091 5180-4124 5181-1291 5180-4161 0100-0542 5180-4160 0100-2144 VLZT1C 5180-4131 5080-8750 5180-4103 G3452-60570 5080-8750 0100-0071 0100-0091 5180-4103 G3452-60570 5180-4129 5181-1291 0100-0124
AG0100-0071EA 32.13 USD InStock
AG0100-0071 AG5080-8751 AG5080-8750 CAAG0100-0071 CA5180-4131 AGG1544-20590 CA0100-0091 CA0100-0090 CAAGVLZT1C CA5180-4105 AG0100-0161 CAG1544-20590 CA0100-0542 AG5180-4121 AG0100-0088 CAG3452-60570 AG5180-4160 AG0100-2144 CA5180-4103 AG5180-4161 AG0100-0124 AG0100-0128 AG0100-0969 AG5180-4127 AG5180-4128 AG5180-4129 AG0100-0091 AG5180-4124 AG0100-0090 CA5061-3303 AG5180-4131 CA5181-1291 AG0100-0053 AG5061-3303 AGG3452-60570 CA5080-8751 CA5080-8750 CAPL1310-0003 CAAG0100-0088 CA5180-4127 CA5180-4129 CA0100-0124 CA5180-4128 AGPL1310-0003 CA0100-0128 CA5180-4124 AGVLZT1C CA0100-0969 CA5180-4121 AG0100-0542 CA5180-4161 CA5180-4160 CAAGAG01000053 AG5181-1291 AG5180-4105 CA0100-0161 CA0100-2144 AG5180-4103
CrossLab GC Fittings, Agilent Technologies
Fittings and Connectors
GC fittings and GC nuts in brass, stainless steel and UltiMetal Plus, in a variety of sizes.
  • Designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent GC fittings and GC nuts for instrument plumbing are available in brass, stainless steel, and UltiMetal Plus, and in sizes ranging from ¹/₁₆ to ¹/₄".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty