Would you like to log in again?
CrossLab Graphite/Vespel Ferrules for GC, Agilent Technologies
Supplier: Agilent Technologies
5062-3538 CR213200 500-1004 5181-3323 5062-3581 5181-3388 0100-1332 5062-3512 0100-1332 CP4784 5062-3538 5181-3308 5062-3514 5062-3580 5062-3512 5062-3514 5181-3308 5181-3388 5062-3581 5062-3508 CR213200 5062-3580 5181-3323 CP4784 500-1004 5062-3508
AG500-1004PK 53.45 USD InStock
AG500-1004 CAAGCP4784 CA5062-3538 AGCP4784 CAAG500-1004 CA76514-662 CA5181-3308 CA76508-056 AGCR213200 AG5062-3512 AG5062-3514 AG5062-3538 AG5062-3580 AG5062-3581 AG0100-1332 CA5062-3580 CA5062-3581 CAAGCR213200 CA76508-092 CAAG5181-3388 AG5181-3388 AG5062-3508 CA76514-660 AG5181-3323 AG5181-3308 CA0100-1332
CrossLab Graphite/Vespel Ferrules for GC, Agilent Technologies
Fittings and Connectors
Graphite/Vespel (15%/85%) ferrules, available in different configurations, provide leak-free connections between the column and injector on your GC.
  • Composed of 85% Vespel polyimide and 15% graphite
  • Non-porous material makes them ideal for MS applications
  • Withstand temperatures as high as 350°C
  • Can be reused and reinstalled due their resistance to deformation
These ferrules are ideal for MS applications as their nonporous material prohibits oxygen contamination, and they can withstand temperatures as high as 350°C. They are also widely used in GC inlets and detectors as they can be reused and reinstalled due their resistance to deformation. Agilent offers a comprehensive selection of graphite-Vespel ferrules for Gas Chromatography and Mass Spectrometry. Our diverse portfolio ensures you will find the correct sized ferrule ID for your GC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty