Would you like to log in again?
CrossLab Sampler and Maintenance Kits for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
G7157-68711 G1329-68727 5063-6506 G4216-68711 G4216-68711 G1329-68727 5063-6506 G7157-68711
AGG1329-68727KT 61.33 USD InStock
CAG1329-68727 CA5063-6506 AGG1329-68727 AG5063-6506 CAAGG715768711 AGG4216-68711 AGG7157-68711 CAAGG421668711
CrossLab Sampler and Maintenance Kits for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A comprehensive range of maintenance, accessory, and upgrade kits for HPLC samplers.
  • Noisy baseline, shorter column life, lower analytical sensitivity, and variable retention times can be caused by a lack of preventive maintenance
  • HPLC autosamplers can work at their best with the Agilent preventive maintenance kits
  • Routine sampler maintenance should be done on a regular basis to keep your Agilent LC system performing at its optimum
Agilent provides a comprehensive range of kits for HPLC samplers. Maintenance kits include supplies that require exchanging on a regular basis, accessory kits contain parts to always have on-hand if needed, and upgrade kits enable modification of the original setup.
All parts are manufactured to Agilent's highest levels of quality and have been tested on Agilent instruments to ensure performance that will significantly increase your analytical, instrument, and laboratory efficiency.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty