Fittings made of different materials to ensure reliable sealing and optimal performance in your LC.
- Innovative InfinityLab Quick Connect and Quick Turn fittings providing finger-tight, reusable, and zero-dead-volume connections
- A variety of fitting materials are available, including stainless steel, PEEK, and polyketone, for different pressure ranges and positions
- Different dimensions covering Swagelok-type or metric M4/M3-type connection ports
- An array of nuts, ferrules, various color identifiers, and other supplies are also available for all fitting types
Agilent has created a large variety of fittings made of different materials to ensure the ongoing security and performance of your LC equipment. Utilizing the correct fitting style and format, as specified by the manufacturer, is important for ensuring accuracy and robust performance in every aspect of your apparatus.;InfinityLab Quick Connect fittings offer truly finger-tight connections up to 1300 bar, for column connections as easy as pressing a lever. For harder-to-reach areas of the LC instrument, try InfinityLab Quick Turn fittings. You can find the right capillaries quickly and easily using our online selection tool.