Would you like to log in again?
Sampler Cones and Care Packages for ICP-MS
Supplier: Agilent
CrossLab
5067-0294 G3280-67036 5067-0294 G3280-67056 G3280-67056 G3280-67036 G1820-65239 G3280-67040 G3280-67061 5067-0295 5067-0295 G1820-65239 G3280-67061 G1820-65238 G1820-65238 G3280-67142 G3280-67142 G3280-67040
AG5067-0294KT 1650.57 USD InStock
CAG3280-67061 CAG1820-65239 CAG3280-67040 CAG1820-65238 AG5067-0294 AG5067-0295 AGG1820-65239 AGG1820-65238 CAAGG328067036 AGG3280-67061 AGG3280-67040 CAAGG328067142 AGG3280-67142 CAAGG328067056 AGG3280-67056 AGG3280-67036 CAAG5067-0294 CAAG5067-0295
Sampler Cones and Care Packages for ICP-MS
Chromatography Supplies Chromatography Spare Parts
Agilent sampler cones and cone care packages ensure ICP-MS system stability and long lifetime.
  • Nickel tipped sampler cones are suitable for most common applications and provide the most economical operation
  • Nickel tipped, nickel plated sampler cones are recommended for samples containing >0.5% HCl, or for routine operation with (U)HMI
  • Platinum tipped sampler cones are required for analysis of aggressive acids (especially HF) and when O₂/Ar option gas is used for analysis of organic solvents
  • Platinum tipped, nickel plated sampler cones are recommended for analysis of aggressive acids (especially HCl, HF, and HClO₄) and for higher-matrix samples
  • Platinum tipped sampler cones with large 18 mm insert are recommended for for high-viscosity and high-boiling-point acids (e.g. H2SO₄, H3PO₄), or for the lowest LODs with semiconductor applications
  • Cone care kits are also available for all Agilent ICP-MS systems, which bundle all the supplies you need to replace and maintain your interface cones in one convenient package
Agilent nickel or platinum tipped sampler cones use high-purity materials to minimize background and ensure stability within the plasma. Cones are typically the biggest supplies cost in routine ICP-MS operation. Protect your investment and ensure ease of use by choosing genuine Agilent cones.
High-quality interface cones are key to the sensitivity and stability of an ICP-MS. Our sampler cones are manufactured to stringent specifications and rigorously tested to ensure excellent batch to batch reproducibility. The orifice dimensions and tip geometry are optimized and tightly controlled.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty