Would you like to log in again?
CrossLab Capillary and Tubing Kits for HPLC, Agilent Technologies
Supplier: Agilent
5067-4250 5067-6697 5067-6804 5067-6801 5067-4294 5067-4270 5004-0003 5067-6802 5067-6802 5061-3304 5065-9939 5067-6708 5067-6800 5067-4300 5067-4234 5067-6187 5061-3315 5067-4249 5067-4270 5067-6598 5067-6614 5065-9938 5065-9939 5067-4633 5065-9937 5067-6615 5065-9937 5013-0002 5067-6598 5067-4249 5067-4251 G1316-68716 5067-4633 5067-4252 5065-9938 5067-6187 5067-4251 5067-6596 5067-4250 G5611-68711 G5611-68711 5067-6615 5067-6707 5067-6181 5067-4252 5061-3315 5004-0003 5067-6596 5067-6708 5067-6597 G1316-68716 5013-0002 5067-6597 5067-6614 5067-6181 5067-6697 5061-3304 5067-4300 5067-4294 5067-6800 5067-4234
AG5004-0003KT 152.38 USD InStock
AG5004-0003 76537-448 CAAG5067-4294 AG5013-0002 CAAG5067-4250 CAAG5067-4251 AG5067-4250 AG5067-4294 AG5067-4251 AG5067-4252 AG5067-6596 AG5065-9937 AG5067-6598 AG5067-6597 AG5065-9939 AGG1316-68716 CAAG5065-9938 AG5065-9938 CAAG5067-6598 CAAG5004-0003 CAAG5067-6597 CAAG5067-6596 CAAG5067-4252 AG5067-6804 CAAG5067-6708 CAAG5067-4249 CAAGG131668716 CAAG5013-0002 CAAG5061-3315 CAAG5067-6187 CAAG5067-6181 AG5067-4300 AG5067-6800 AG5067-6802 CA5061-3304 AG5067-6801 CA5065-9939 CA5065-9937 AG5061-3304 CAAG5067-6615 CAAG5067-6614 AGG5611-68711 AG5067-6181 AG5067-4270 CAAG5067-4270 AG5067-6697 CAAG5067-4633 AG5067-4234 CAAG5067-4234 CAAG5067-6697 AG5067-4633 AG5067-6615 AG5067-6614 AG5067-6708 CAAG5067-6802 AG5061-3315 CAAGG561168711 AG5067-6187 CAAG5067-6800 CAAG5067-4300 AG5067-4249
CrossLab Capillary and Tubing Kits for HPLC, Agilent Technologies
Tubing Chromatography Tubing
A complete set of system capillaries and fittings to efficiently ramp up usage of LC instruments.
  • Carefully selected capillary dimensions (inner diameter and length) to minimize extra-column volume and optimize separation efficiency
  • Peace of mind of having everything needed to make necessary connections on your system
  • InfinityLab Quick Connect and Quick Turn fittings in the InfinityLab capillary kits ensure ease of use and zero dead-volume column connection
Agilent InfinityLab capillary kits are complete sets of system capillaries, including InfinityLab Quick-Connect and Quick Turn fittings, to efficienctly ramp up usage of InfinityLab LC Series instruments.
Agilent capillary/fitting starter kits contain the most widely used connection capillaries, fittings, and tools needed for older LC systems. Capillary dimensions are optimized to minimize extra-column volume to ensure highest separation efficiency.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty