Would you like to log in again?
CrossLab Seal Wash Supplies for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
5065-9952 0905-1718 5065-9952 G9203-80004 5067-6131 5001-3743 5062-2484 01018-60027 5067-6131 0905-1718 5042-8954 G5611-26210 G9203-80004 5001-3743 5042-8507 5065-9978 5067-4793 0905-1175 G5611-26210 5065-9978 5067-4793 01018-60027 0905-1175 5062-2484 G4220-63010 5042-8507 G4220-63010 5042-8954
AGG4220-63010EA 196.23 USD InStock
AGG4220-63010 AG5065-9952 CAAG5065-9952 CA76514-528 AG5065-9978 AG5062-2484 AGG5611-26210 CA5065-9978 CAAG5067-6131 AG0905-1718 CA76508-054 AG5042-8954 CA5042-8507 AG5067-6131 AGG9203-80004 AG5067-4793 CAAGG422063010 CAAG5067-4793 AG01018-60027 AG5001-3743 CAAGG561126210 AG0905-1175 CA5001-3743 CA0905-1718 AG5042-8507 CAAGG920380004 CAAG5042-8954 CA01018-60027
CrossLab Seal Wash Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Seal wash supplies help increase seal lifetime when highly concentrated buffer solutions are used.
  • Water/isopropanol (90/10) is recommended as the wash solvent
  • Wash the flow path with clear solvent after using concentrated buffers to increase the piston and seal lifetime
  • Routine pump maintenance should be done on a regular basis to keep your Agilent LC system performing at its optimum
Agilent seal wash kits include all supplies needed to modify the pump head to allow flushing the back of the seal with a wash solvent. The routine use of highly concentrated buffer solutions (100 mM) will reduce the life of seals and pistons in your pump. The use of seal wash in HPLC systems helps to increase the performance and lifetime of the piston seals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty