Would you like to log in again?
CrossLab Degasser Supplies for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
0100-1710 9301-1337 5062-2483 5063-6599 G1322-67300 5062-2483 5063-6598 5067-5388 5067-5380 5067-5378 5022-2155 5042-8922 5067-5387 5022-2155 5067-5383 G1379-67310 5067-6624 8710-1930 5067-5383 G1322-68705 5067-5380 5067-5388 5067-4798 5067-6624 5063-6599 5067-4798 G1322-68705 G1322-67300 9301-1337 8710-1930 5042-8922 5063-6598 0100-1710 5067-5378 5067-5387 G1379-67310
AG5022-2155PK 67.77 USD InStock
CAG1322-67300 AG5022-2155 AGG1322-68705 AG5062-2483 CA5022-2155 AG5067-5380 CAAG5067-5383 CA76508-052 CA8710-1930 CA5067-5378 AG5067-5383 CAG1379-67310 CAAG5067-5380 AG5067-5388 CA5063-6599 CA5042-8922 AG5067-5387 CA5063-6598 CAAG5067-5387 CAAG5067-5388 AG5067-4798 CA0100-1710 AGG1379-67310 CA9301-1337 AG0100-1710 CAG1322-68705 AG5063-6599 AG9301-1337 AG5042-8922 AG5063-6598 CA5067-4798 CAAG5067-6624 AG5067-5378 AG5067-6624 AG8710-1930 AGG1322-67300
CrossLab Degasser Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Extensive range of supplies for degasser instruments to remove dissolved oxygen from HPLC solvents.
  • Fast and easy flushing, purging, priming, and solvent exchange;High degassing efficiency for the most demanding applications
  • Highly reproducible analyses
  • Very low baseline noise
Agilent provides an extensive range of supplies for degasser instruments to optimize the HPLC results by removing any trace of dissolved oxygen in HPLC solvents. The Agilent vacuum degasser is recommended for maximum sensitivity in the low-UV wavelength range, for high injection precision, for high retention time reproducibility, and for low flow rate methods.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty