Extensive range of supplies for degasser instruments to remove dissolved oxygen from HPLC solvents.
- Fast and easy flushing, purging, priming, and solvent exchange;High degassing efficiency for the most demanding applications
- Highly reproducible analyses
- Very low baseline noise
Agilent provides an extensive range of supplies for degasser instruments to optimize the HPLC results by removing any trace of dissolved oxygen in HPLC solvents. The Agilent vacuum degasser is recommended for maximum sensitivity in the low-UV wavelength range, for high injection precision, for high retention time reproducibility, and for low flow rate methods.