Would you like to log in again?
CrossLab Pump and Maintenance Kits for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
G1311-68711 G1312-68711 5067-6137 G4220-20001 G1312-68711 G4220-20001 G1311-68711 5067-6137
AGG4220-20001EA 52.55 USD InStock
AGG4220-20001 AGG1312-68711 AG5067-6137 CAAGG422020001 AGG1311-68711 CAAG5067-6137 CAAGG131268711 CAAGG131168711
CrossLab Pump and Maintenance Kits for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A comprehensive range of kits for maintaining and upgrading HPLC pumps.
  • A lack of preventive maintenance can cause noisy baselines, shorter column life, lower analytical sensitivity, and variable retention times
  • HPLC pumps work at their best with the Agilent preventive maintenance kits
  • Routine pump maintenance should be done on a regular basis to keep your Agilent LC system performing at its optimum
  • By following a regular maintenance routine you can count on maximum uptime and a steady, accurate solvent flow for the life of the pump
Agilent provides a comprehensive range of kits for HPLC pumps. Maintenance kits include the right supplies to be exchange on a regular basis. Accessory kits contain parts to always have on-hand. Upgrade kits allow users to change the original setup.
All parts are manufactured to Agilent's highest levels of quality and have been tested in Agilent instruments to ensure they work perfectly with the rest of the system.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty