Would you like to log in again?
CrossLab Column Thermostat Accessories for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
G1316-87303 5062-8588 G1316-87303 5043-1465 5067-5917 5067-6654 G7116-68004 5062-8588 G7163-60002 G7116-68003 G7116-68003 5067-6849 5001-3702 5067-6849 5067-5963 5067-6654 G7116-60013 G7116-60013 G1316-60001 G1316-60001 G7116-68004 5067-5963 5063-6526 5067-5917 5043-1465 G7163-60002 5001-3702 5063-6526
AG5062-8588PK 196.23 USD InStock
AG5062-8588 AG5063-6526 CA5062-8588 CAAG5067-5963 AGG7116-68003 CAAG5067-6654 AGG7116-68004 AG5067-6654 CAAGG711660013 AG5067-5963 CAAGG131660001 AG5043-1465 CAAG5001-3702 CAAG5067-5917 CAAG5067-6849 AG5067-6849 CAAG5043-1465 AG5067-5917 AGG7163-60002 AGG7116-60013 CAAGG716360002 CA5063-6526 CAG7116-68004 CAG7116-68003 AGG1316-87303 CA76515-212 AGG1316-60001 AG5001-3702
CrossLab Column Thermostat Accessories for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Supplies to ensure the performance of the thermostatted column compartment in HPLC systems.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Column Int.Ø tags allow you to track various column properties and usage parameter including but not limited to column identity, lot and batch number, the last injection date, number of injections, and the max temperature used
  • Column Int.Ø tags ensure traceability of analysis, documenting columns and conditions for routine analysis easy and adding confidence to any analysis
Agilent provides a comprehensive range of high-quality column thermostat accessories and supplies, including column tags and heat exchangers, to support your efforts in increasing your analytical results.
All parts are manufactured to Agilent's highest levels of quality and have been tested in Agilent instruments to ensure they work perfectly with the rest of the system.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty